4-(Trifluoromethyl)benzene-1,2-diol 97%
Catalog # APOSPC28227-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:178.109 g/mol
Storage Temperature:Ambient
CAS Number:37519-09-8
Formula:C₇H₅F₃O₂
MDL Number:MFCD18395730