4-Fluoro-3-oxo-1-phenyl-2,3-dihydro-1H-indene-1-carboxylic acid, tech
Catalog # APOSPC28137-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:270.255 g/mol
Storage Temperature:Ambient
CAS Number:339010-41-2
Formula:C₁₆H₁₁FO₃
MDL Number:MFCD00232260