Order Entry
Ireland
ContactUsLinkComponent
1-Methyl-2-oxo-5-[3-(trifluoromethyl)phenyl]-1,2-dihydropyridine-3-carbonitrile, tech
1-Methyl-2-oxo-5-[3-(trifluoromethyl)phenyl]-1,2-dihydropyridine-3-carbonitrile, tech
Catalog # APOSPC28117-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Methyl-2-oxo-5-[3-(trifluoromethyl)phenyl]-1,2-dihydropyridine-3-carbonitrile, tech
Catalog # APOSPC28117-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :278.229 g/mol
Storage Temperature:Ambient
CAS Number:338964-42-4
Formula:C₁₄H₉F₃N₂O
MDL Number:MFCD00232375