Order Entry
Ireland
ContactUsLinkComponent
2,2,2-Trifluoro-N-(4-methoxy-3-oxo-2,3-dihydro-1H-inden-1-yl)acetamide, tech
2,2,2-Trifluoro-N-(4-methoxy-3-oxo-2,3-dihydro-1H-inden-1-yl)acetamide, tech
Catalog # APOSPC28035-1G
Supplier:  Apollo Scientific
CAS Number:  
2,2,2-Trifluoro-N-(4-methoxy-3-oxo-2,3-dihydro-1H-inden-1-yl)acetamide, tech
Catalog # APOSPC28035-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :273.208 g/mol
Storage Temperature:Ambient
CAS Number:861209-50-9
Formula:C₁₂H₁₀F₃NO₃
MDL Number:MFCD03102804