5-Methoxy-2-[3-(trifluoromethyl)phenyl]pyrimidin-4-ol, tech
Catalog # APOSPC28016-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:270.207 g/mol
Storage Temperature:Ambient
CAS Number:860788-30-3
Formula:C₁₂H₉F₃N₂O₂
MDL Number:MFCD04124708