2-Bromo-4-(methoxymethyl)fluorobenzene 95+%
Catalog # APOSPC2422-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:219.051 g/mol
Storage Temperature:Ambient
CAS Number:887268-22-6
Formula:C₈H₈BrFO
MDL Number:MFCD04116324