Cyclopropylmethyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate 97%
Catalog # APOSPC2115-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:280.631 g/mol
Boiling Pt:108…110 °C (0,5 mm Hg)
:GHS07
Storage Temperature:Ambient
CAS Number:175137-31-2
Formula:C₁₀H₈ClF₃N₂O₂
MDL Number:MFCD00068151