[6-(trifluoromethyl)pyridin-3-yl]methyl 6-(trifluoromethyl)pyridine-3-carboxylate
Catalog # APOSPC210099-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:350.216 g/mol
Melting Pt:102…104 °C
Storage Temperature:Ambient
CAS Number:1781241-48-2
Formula:C₁₄H₈F₆N₂O₂
MDL Number:MFCD28806001