4-Bromo-5-fluoro-2-methylbenzoic acid 98%
Catalog # APOSPC201125-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:233.034 g/mol
Storage Temperature:Ambient
CAS Number:1349715-55-4
Formula:C₈H₆BrFO₂
MDL Number:MFCD18917157