5-Bromo-4-fluoro-2,3-dihydro-1H-inden-1-one 96%
Catalog # APOSPC201110-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:229.046 g/mol
Storage Temperature:Ambient
CAS Number:127425-74-5
Formula:C₉H₆BrFO
MDL Number:MFCD18208171