(S)-5-Fluoro-2,3-dihydro-1H-inden-1-amine hydrochloride 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1114333-11-7
:2103399-35-3
Formula:C₉H₁₀FN·ClH
MDL Number:MFCD16295141
:187.642 g/mol
Storage Temperature:Ambient
Frequently Bought Together