2-Bromo-3-(trifluoromethyl)-2-butenoic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:885276-60-8
:
Formula:C₅H₄BrF₃O₂
MDL Number:MFCD09998065
:232.983 g/mol
Melting Pt:72…74 °C
Storage Temperature:Ambient
Frequently Bought Together