3,5-Dimethyl-2,4,6-trifluorobenzoyl chloride
Catalog # APOSPC10084-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:222.592 g/mol
Storage Temperature:Ambient
UN:1760
ADR:8,III
:GHS05
CAS Number:886762-25-0
Formula:C₉H₆ClF₃O
MDL Number:MFCD04039201