Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Perfluoro-4-methylbenzoic acid
Perfluoro-4-methylbenzoic acid
Perfluoro-4-methylbenzoic acid
APOSPC0506-5G
:
Apollo Scientific
:
Perfluoro-4-methylbenzoic acid
APOSPC0506-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
262.081 g/mol
:
109…110 °C
:
GHS07
:
Ambient
:
5216-22-8
:
C₈HF₇O₂
:
MFCD01631500