4,7-Difluoroindan-1-one
Catalog # APOSPC01482-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:168.14 g/mol
Storage Temperature:Ambient
CAS Number:130408-16-1
Formula:C₉H₆F₂O
MDL Number:MFCD07772126