Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(2-methylpyridin-3-yl)methanol
(2-methylpyridin-3-yl)methanol 95%
(2-methylpyridin-3-yl)methanol 95%
APOSOR959578-1G
:
Apollo Scientific
:
(2-methylpyridin-3-yl)methanol 95%
APOSOR959578-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
123.152 g/mol
:
Ambient
:
56826-61-0
:
C₇H₉NO
:
MFCD00030820