Order Entry
Ireland
ContactUsLinkComponent
 
Propyl-3-hydroxybenzoate 95%
Catalog # APOSOR957605-1G
Supplier:  Apollo Scientific
CAS Number:  
Propyl-3-hydroxybenzoate 95%
Catalog # APOSOR957605-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :180.2 g/mol
Storage Temperature:Ambient
CAS Number:38567-05-4
Formula:C₁₀H₁₂O₃
MDL Number:MFCD03424456