Order Entry
Ireland
ContactUsLinkComponent
2-Chloro-5-(hydroxymethyl)phenol 96%
2-Chloro-5-(hydroxymethyl)phenol 96%
Catalog # APOSOR957467-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-5-(hydroxymethyl)phenol 96%
Catalog # APOSOR957467-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :158.582 g/mol
Storage Temperature:Ambient
CAS Number:1261598-26-8
Formula:C₇H₇ClO₂
MDL Number:MFCD18393156