Order Entry
Ireland
ContactUsLinkComponent
1-Chloro-3-methoxy-5-methylbenzene 95%
1-Chloro-3-methoxy-5-methylbenzene 95%
Catalog # APOSOR956991-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Chloro-3-methoxy-5-methylbenzene 95%
Catalog # APOSOR956991-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :156.609 g/mol
Storage Temperature:Ambient
CAS Number:82477-66-5
Formula:C₈H₉ClO
MDL Number:MFCD08234651