Order Entry
Ireland
ContactUsLinkComponent
Methyl (2S,3aS,7aS)-octahydro-1H-indole-2-carboxylate 96%
Methyl (2S,3aS,7aS)-octahydro-1H-indole-2-carboxylate 96%
Catalog # APOSOR955505-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl (2S,3aS,7aS)-octahydro-1H-indole-2-carboxylate 96%
Catalog # APOSOR955505-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :183.247 g/mol
Storage Temperature:Ambient
CAS Number:192436-84-3
Formula:C₁₀H₁₇NO₂
MDL Number:MFCD20662144