
4-Chloro-6-methyl-1H-pyrrolo[3,2-c]pyridine 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:178268-92-3
:
Formula:C₈H₇ClN₂
MDL Number:MFCD12401559
:166.608 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together