3-Allyl-4-methoxybenzaldehyde 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:67483-48-1
:
Formula:C₁₁H₁₂O₂
MDL Number:MFCD08691767
:176.212 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together