Order Entry
Ireland
ContactUsLinkComponent
(1S,2S)-N1,N1-Dimethylcyclohexane-1,2-diamine 95%
(1S,2S)-N1,N1-Dimethylcyclohexane-1,2-diamine 95%
Catalog # APOSOR930741-5G
Supplier:  Apollo Scientific
CAS Number:  
(1S,2S)-N1,N1-Dimethylcyclohexane-1,2-diamine 95%
Catalog # APOSOR930741-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

About this item

 :
 :142.242 g/mol
Storage Temperature:Ambient
CAS Number:894493-95-9
Formula:C₈H₁₈N₂
MDL Number:MFCD09952017