5-Methyl-[1,2,4]triazolo[1,5-a]pyridin-2-ylamine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1214901-53-7
:
Formula:C₇H₈N₄
MDL Number:MFCD11846608
:148.165 g/mol
Storage Temperature:Ambient
Frequently Bought Together