2,8-Dimethyl[1,2,4]triazolo[1,5-a]pyridin-6-ylamine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1417637-80-9
:
Formula:C₈H₁₀N₄
MDL Number:MFCD26593827
:162.192 g/mol
Storage Temperature:Ambient
Frequently Bought Together