(6-Chloropyridazin-3-yl)methanol
Catalog # APOSOR921872-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:144.559 g/mol
Storage Temperature:Ambient
CAS Number:1011487-94-7
Formula:C₅H₅ClN₂O
MDL Number:MFCD11977451