5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-inden-2-ol 98%
Catalog # APOSOR915673-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:260.136 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1857348-77-6
Formula:C₁₅H₂₁BO₃
MDL Number:MFCD27936516