4-(4-Chlorophenyl)-1,3-thiazole 98%
Catalog # APOSOR906578-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:195.669 g/mol
Storage Temperature:Ambient
CAS Number:1826-22-8
Formula:C₉H₆ClNS
MDL Number:MFCD02665933