5-Chloro-3-methylpyridine-2-carbonitrile
Catalog # APOSOR60143-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:152.581 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:156072-84-3
Formula:C₇H₅ClN₂
MDL Number:MFCD17214209