
6'-Bromo-2'-hydroxyacetophenone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:55736-69-1
:
Formula:C₈H₇BrO₂
MDL Number:MFCD13193630
:215.044 g/mol
Melting Pt:116…119,2 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together