Order Entry
Ireland
ContactUsLinkComponent
(7-Chloro-[1,2,4]triazolo[1,5-a]pyridin-2-yl)methanol ≥98%
(7-Chloro-[1,2,4]triazolo[1,5-a]pyridin-2-yl)methanol ≥98%
Catalog # APOSOR510172-100MG
Supplier:  Apollo Scientific
CAS Number:  
(7-Chloro-[1,2,4]triazolo[1,5-a]pyridin-2-yl)methanol ≥98%
Catalog # APOSOR510172-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :183.595 g/mol
Storage Temperature:Ambient
CAS Number:1896916-30-5
Formula:C₇H₆ClN₃O