7-Bromo-2,3-dihydro-1H-inden-1-ol ≥97%
Catalog # APOSOR510033-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:213.071 g/mol
Storage Temperature:Ambient
CAS Number:1196049-18-9
Formula:C₉H₉BrO
MDL Number:MFCD24843663