Order Entry
Ireland
ContactUsLinkComponent
(R)-(4-Methylmorpholin-3-yl)methanol ≥97%
(R)-(4-Methylmorpholin-3-yl)methanol ≥97%
Catalog # APOSOR510000-250MG
Supplier:  Apollo Scientific
CAS Number:  
(R)-(4-Methylmorpholin-3-yl)methanol ≥97%
Catalog # APOSOR510000-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :131.173 g/mol
Storage Temperature:Refrigerator
UN:1760
ADR:8,III
CAS Number:1620510-51-1
Formula:C₆H₁₃NO₂
MDL Number:MFCD26792468