Methyl[methyl(oxo)phenyl-lambda6-sulphanylidene]amine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:30004-67-2
:
Formula:C₈H₁₁NOS
MDL Number:MFCD00051932
:169.244 g/mol
Storage Temperature:Ambient
Frequently Bought Together