3-(2-Acetamidoethyl)-1H-indol-5-yl acetate
Catalog # APOSOR4891-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
Melting Pt:121 °C
:260.288 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:28026-16-6
Formula:C₁₄H₁₆N₂O₃
MDL Number:MFCD00209948