Order Entry
Ireland
ContactUsLinkComponent
1-Methyl-DL-tryptophan 95%
1-Methyl-DL-tryptophan 95%
Catalog # APOSOR480297-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Methyl-DL-tryptophan 95%
Catalog # APOSOR480297-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :218.252 g/mol
Storage Temperature:Ambient
CAS Number:26988-72-7
Formula:C₁₂H₁₄N₂O₂
MDL Number:MFCD00005803