
4,6-Dichloro-3-methylpyridazine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:68240-43-7
:
Formula:C₅H₄Cl₂N₂
MDL Number:MFCD21604745
:163.005 g/mol
Melting Pt:35…37 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together