N-[1-(2-Bromophenyl)ethyl]cyclopropanamine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2367002-83-1
:
Formula:C₁₁H₁₄BrN·ClH
MDL Number:MFCD31695694
:276.601 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together