N,N-Dioctylbenzimidazole-5-carboxamide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2006277-84-3
:
Formula:C₂₄H₃₉N₃O
MDL Number:MFCD28505587
Molecular Weight:385.586 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together