4-Cbz-piperazine-1-carbonyl chloride
Catalog # APOSOR471201-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:282.723 g/mol
Storage Temperature:Ambient
CAS Number:25539-27-9
Formula:C₁₃H₁₅ClN₂O₃
MDL Number:MFCD09743772