4-(Cyclopropylmethoxy)-3-methylbenzoic acid
Catalog # APOSOR470794-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:206.238 g/mol
Storage Temperature:Ambient
CAS Number:1252903-16-4
Formula:C₁₂H₁₄O₃
MDL Number:MFCD16852894