6-[Bis(Boc)amino]purine
Catalog # APOSOR470783-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:335.358 g/mol
Storage Temperature:Ambient
CAS Number:309947-86-2
Formula:C₁₅H₂₁N₅O₄
MDL Number:MFCD17676144