Order Entry
Ireland
ContactUsLinkComponent
3-Formyl-4-methylbenzoic acid
3-Formyl-4-methylbenzoic acid
Catalog # APOSOR470735-5G
Supplier:  Apollo Scientific
CAS Number:  
3-Formyl-4-methylbenzoic acid
Catalog # APOSOR470735-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :164.158 g/mol
Storage Temperature:Ambient
CAS Number:69526-89-2
Formula:C₉H₈O₃
MDL Number:MFCD20660432