Ethyl 3-Methyl-2-oxo-3-butenoate
Catalog # APOSOR470596-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:142.152 g/mol
Storage Temperature:Ambient
CAS Number:50331-71-0
Formula:C₇H₁₀O₃
MDL Number:MFCD23381118