5-Propylpyrazole-3-carboxamide
Catalog # APOSOR470503-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
:
:153.182 g/mol
Storage Temperature:Ambient
CAS Number:957129-48-5
Formula:C₇H₁₁N₃O
MDL Number:MFCD16619745