Order Entry
Ireland
ContactUsLinkComponent
Ethyl 2-Oxo-5-phenyl-2,3-dihydro-1H-imidazole-4-carboxylate
Ethyl 2-Oxo-5-phenyl-2,3-dihydro-1H-imidazole-4-carboxylate
Catalog # APOSOR470450-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 2-Oxo-5-phenyl-2,3-dihydro-1H-imidazole-4-carboxylate
Catalog # APOSOR470450-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :232.235 g/mol
Storage Temperature:Ambient
CAS Number:92809-78-4
Formula:C₁₂H₁₂N₂O₃
MDL Number:MFCD22574973