4-(Boc-amino)-1-(6-chloro-3-pyridazinyl)piperidine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:596817-46-8
:
Formula:C₁₄H₂₁ClN₄O₂
MDL Number:MFCD09607516
:312.795 g/mol
Storage Temperature:Ambient
Frequently Bought Together