(2S)-N1,N1,3-Trimethyl-1,2-butanediamine dihydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:847696-16-6
:
Formula:C₇H₁₈N₂·2ClH
MDL Number:MFCD31560222
Molecular Weight:203.153 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together