
(S)-2-(Boc-amino)-2-phenylethylamine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:137102-30-8
:
Formula:C₁₃H₂₀N₂O₂
MDL Number:MFCD09885895
:236.31 g/mol
Melting Pt:85…90 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together