Order Entry
Ireland
ContactUsLinkComponent
 
3-Amino-1-(3-bromophenyl)thiourea
Catalog # APOSOR450118-250MG
Supplier:  Apollo Scientific
CAS Number:  
3-Amino-1-(3-bromophenyl)thiourea
Catalog # APOSOR450118-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :246.128 g/mol
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
CAS Number:116567-17-0
Formula:C₇H₈BrN₃S
MDL Number:MFCD00041285